missing translation for 'onlineSavingsMsg'
Learn More

Spermidine Trihydrochloride, Fisher BioReagents

Product Code. 10348084
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348084 25 g Amber Glass 25g
11423084 1 g Amber Glass 1g
11433084 5 g Amber Glass 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10348084 Supplier Fisher BioReagents Supplier No. BP254025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms

Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins

  • Spermidine is an aliphatic polyamine used in various biochemical applications due to its involvement in many metabolic functions within organisms
  • Forms stable compounds with nucleic acids and may be used for the purification of DNA-binding proteins
  • Also serves as a plant growth regulator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334-50-9
Molecular Formula C7H22Cl3N3
Molecular Weight (g/mol) 254.62
MDL Number MFCD00012918
InChI Key LCNBIHVSOPXFMR-UHFFFAOYSA-N
Synonym spermidine trihydrochloride, n-3-aminopropyl-1,4-butanediamine trihydrochloride, spermidine hydrochloride, n1-3-aminopropyl butane-1,4-diamine trihydrochloride, unii-1o14bed398, 1,4-butanediamine, n-3-aminopropyl-, trihydrochloride, spermidine, trihydrochloride, 1,4-butanediamine, n-3-aminopropyl , hydrochloride, n-3-aminopropyl butane-1,4-diamine trihydrochloride, 4-azoniaoctamethylenediammonium trichloride
PubChem CID 9539
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine;trihydrochloride
SMILES [H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN

Specifications

Melting Point 248°C
Color White
pH 3 to 6
Assay Percent Range ≥99 %
Packaging Amber Glass
Quantity 25 g
Merck Index 15, 8869
Formula Weight 254.63
Percent Purity 99% min.
Physical Form Solid
Chemical Name or Material Spermidine Trihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Spermidine trihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.