missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, MedChemExpress

Product Code. 16488806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488806 Supplier Medchem Express Supplier No. HYB1776100mg
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Spermidine maintains cell membrane stability, increases antioxidant enzymes activities, improving photosystem II (PSII), and relevant gene expression. Spermidine significantly decreases the H2O2 and O2 contents.

Spermidine maintains cell membrane stability, increases antioxidant enzymes activities, improving photosystem II (PSII), and relevant gene expression. Spermidine significantly decreases the H2O2 and O2 contents.

Specifications

Chemical Name or Material Spermidine
CAS 124-20-9
Color Colorless
Purity Grade Notes Research
Molecular Formula C7H19N3
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (688.47 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (114.77 mM; ultrasonic and warming and adjust pH to 7 with HCl and heat to 80°C)
SMILES NCCCCNCCCN
Molecular Weight (g/mol) 145.25
Formula Weight 145.25
Percent Purity 97.0%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.