missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, free base, 99.5%, MP Biomedicals™

Product Code. 11474371 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474371 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474371 Supplier MP Biomedicals Supplier No. 0215206801
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008229
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
PubChem CID 1102
ChEBI CHEBI:16610
IUPAC Name (4-aminobutyl)(3-aminopropyl)amine
SMILES NCCCCNCCCN

Specifications

Color Colorless
Density 0.925g/mL
Assay Percent Range ∽ 99%
Quantity 1 g
Formula Weight 145.25
Percent Purity 0.995
Physical Form Liquid
Chemical Name or Material Spermidine free base, Cell Biology and Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.