missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, ∽98%; Molecular biology reagent grade, MP Biomedicals™

Product Code. 11418302 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418302 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11418302 Supplier MP Biomedicals Supplier No. 0219485225
  • ONLINE EXCLUSIVE PRICE
    £790.00
    £740.25 / 25g
    Save £49.75
    6% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 124-20-9
Color Colorless
Assay Percent Range Approx. 99%
Molecular Formula C7H19N3
MDL Number MFCD00008229
Quantity 25 g
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
SMILES NCCCCNCCCN
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine
Molecular Weight (g/mol) 145.25
PubChem CID 1102
ChEBI CHEBI:16610
Formula Weight 145.25
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material Spermidine
Show More Show Less
compliance-icons
Product Identifier
  • Spermidine Free Base
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Spermidine Free Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.