missing translation for 'onlineSavingsMsg'
Learn More

SPEN Antibody, Novus Biologicals™

Product Code. 18718303 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
25µL
Quantity:
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Quantity
18718303 0.10mL -
18456120 25µL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18718303 Supplier Novus Biologicals Supplier No. NBP182952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

SPEN Polyclonal specifically detects SPEN in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Classification Polyclonal
Isotype IgG

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.