missing translation for 'onlineSavingsMsg'
Learn More

SPEN Antibody, Novus Biologicals™

Product Code. 18456120 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18456120 25 μL 25µL
18718303 - 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18456120 Supplier Novus Biologicals Supplier No. NBP18295225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

SPEN Polyclonal specifically detects SPEN in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPEN
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila)
Gene Symbols SPEN
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:NRERAYEHSAYGHHERGTGGFDRTRHYDQDYYRDPRERTLQHGLYYASRSRSPNRFDAHDPRYEPRAREQFTLPSVVHRDIYRDDITREVRGRRPERNYQHSRSRSPHSSQSRNQSPQRLASQASRPTRSPSGSGSRSRSSSSDSI
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23013
Test Specificity Specificity of human SPEN antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.