missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEN Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-58798AF647
This item is not returnable.
View return policy
Description
SPEN Polyclonal antibody specifically detects SPEN in Human samples. It is validated for Western Blot
Specifications
| SPEN | |
| Polyclonal | |
| Western Blot | |
| HIAA0929, KIAA0929nuclear receptor transcription cofactor, MINTMsx2 interacting nuclear target (MINT) homolog, RBM15C, SHARPmsx2-interacting protein, SMART/HDAC1 associated repressor protein, SMART/HDAC1-associated repressor protein, SPEN homolog, spen homolog, transcriptional regulator (Drosophila) | |
| The immunogen recognized by this antibody maps to a region between residue 1350 and 1400 of human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator) using the numbering given in entry NP_055816.2 (GeneID 23013). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23013 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction