missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPEG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553875
This item is not returnable.
View return policy
Description
Immunogen sequence: VYKSVIANKL GKAACYAHLY VTDVVPGPPD GAPQVVAVTG RMVTLTWNPP RSLDMAIDPD SLTYTVQHQV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
Specifications
| SPEG | |
| Polyclonal | |
| Unconjugated | |
| SPEG | |
| aortic preferentially expressed gene 1; aortic preferentially expressed protein 1; APEG1; APEG-1; BPEG; CNM5; KIAA1297; nuclear protein, marker for differentiated aortic smooth muscle and down-regulated with vascular injury; SPEG; SPEG complex locus; SPEGalpha; SPEGbeta; striated muscle preferentially expressed protein kinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10290 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15772 | |
| SPEG | |
| Recombinant protein corresponding to Human SPEG. Recombinant protein control fragment (Product #RP-92276). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction