missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562243
This item is not returnable.
View return policy
Description
Immunogen sequence: RDFSDGVLVA EVIKFYFPKM VEMHNYVPAN SLQQKLSNWG HLNRKVLKRL NFSVPDDVMR KIAQCAPGVV ELVLIPLRQR LEERQRRRKQ GAGSLQELAP QDGSGYMDVG KVAFSISPSR L Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
Microtubule-associated protein involved in the stabilization of microtubules along the axis of migration during radial intercalation. Promotes the establishment and stabilization of an axis of microtubules required for the active migration of cells into the outer epithelium. Microtubule-associated protein that promotes microtubule bundling and stabilizes microtubules against depolymerization in response to cold shock.
Specifications
| SPEF1 | |
| Polyclonal | |
| Unconjugated | |
| SPEF1 | |
| C20orf28; calponin-homology and microtubule-associated protein; CLAMP; SPEF1; SPEF1A; sperm flagellar 1; sperm flagellar protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25876 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y4P9 | |
| SPEF1 | |
| Recombinant protein corresponding to Human SPEF1. Recombinant protein control fragment (Product #RP-88731). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction