missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571342
This item is not returnable.
View return policy
Description
SPEF1 Polyclonal Antibody for Western Blot
Microtubule-associated protein involved in the stabilization of microtubules along the axis of migration during radial intercalation. Promotes the establishment and stabilization of an axis of microtubules required for the active migration of cells into the outer epithelium. Microtubule-associated protein that promotes microtubule bundling and stabilizes microtubules against depolymerization in response to cold shock.
Specifications
| SPEF1 | |
| Polyclonal | |
| Unconjugated | |
| SPEF1 | |
| C20orf28; calponin-homology and microtubule-associated protein; CLAMP; SPEF1; SPEF1A; sperm flagellar 1; sperm flagellar protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25876 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y4P9 | |
| SPEF1 | |
| synthetic peptide directed towards the N-terminal region of human SPEF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction