missing translation for 'onlineSavingsMsg'
Learn More

3M™ Speedglas™ Headband with Airduct 9000

Product Code. 15770008
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
15770008 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15770008 missing translation for 'mfr' 3M™ missing translation for 'supplierNo' 7000144515

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

3M™ Speedglas™ Headband with Airduct 9000 for use with Speedglas™ Welding Helmet Series 9000.

  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Replace worn out or damaged parts
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Speedglas™ Welding Helmet Series 9000
Includes Air duct
Product Type Headband
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.