missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Transfertube™ Tube
Description
- Material: polypropylene
- Volume: 4mL
- 153mm length × 6mm dia.
- I.D.: 16.5mm
- O.D.: 20mm
Specifications
Specifications
| Capacity (Metric) | 4 mL |
| Length (Metric) | 175 mm |
| Material | Polypropylene |
| For Use With | Bacterial or viral colony on agar in microbiology, powders, viscous solutions, slurries, liquids, soil, mud, sludge, core samples |
| Diameter (Metric) | 20 mm |
| Product Type | Transport Tube |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?