missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing

Product Code. 12316903
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
16 mm
20 mm
24 mm
MWCO:
0.1 to 0.5 kDa
0.5 kDa
1 kDa
10 kDa
100 kDa
25 kDa
5 kDa
50 kDa
Unit Size:
10m
Each
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) MWCO unitSize
12316903 10 mm 0.1 to 0.5 kDa Each
11475929 20 mm 0.5 kDa Each
11405939 20 mm 1 kDa Each
11495929 24 mm 0.5 kDa Each
10596734 15 mm 1 kDa Each
11435939 15 mm 5 kDa Each
11445939 20 mm 5 kDa Each
11465939 15 mm 10 kDa Each
11475939 20 mm 10 kDa Each
11435959 6.4 mm 50 kDa Each
11445959 10 mm 50 kDa Each
11455959 15 mm 50 kDa Each
11465959 20 mm 50 kDa Each
11495939 6.4 mm 100 kDa Each
11415949 10 mm 100 kDa Each
11405949 15 mm 100 kDa Each
11425949 20 mm 100 kDa Each
11495949 6.4 mm 25 kDa 10m
11485929 6.4 mm 0.5 kDa Each
11405959 10 mm 25 kDa Each
10353809 16 mm 1 kDa Each
11425939 6.4 mm 5 kDa Each
10069439 10 mm 5 kDa Each
11485939 6.4 mm 10 kDa Each
11455939 10 mm 10 kDa Each
11415959 15 mm 25 kDa Each
11425959 20 mm 25 kDa Each
11415939 6.4 mm 1 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
This item is not returnable. View return policy
Product Code. 12316903 Supplier Spectrum™ Supplier No. 131054
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Selected MWCOs from 100 to 100,000 Daltons

  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution

Order Info

Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.

Specifications

Product Type Spectra/Por Dialysis Membrane Tubing
MWCO 0.1 to 0.5 kDa
Material Cellulose Ester
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Supplied in 0.1% sodium azide solution
Temperature Range (English) 39.2°F to 98.6°F
Temperature Range (Metric) 4°C to 37°C
Quantity Each
For Use With (Application) Ideal for desalting, purification, and molecular separation
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.