missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO

Product Code. 11465919
Change view
Click to view available options
Diameter (Metric):
3.8 mm
7.5 mm
11.5 mm
15 mm
17.8 mm
20.4 mm
21.6 mm
24 mm
25.5 mm
29 mm
32 mm
34.4 mm
45 mm
Quantity:
1 Pk.
Each
MWCO:
1 kDa
10 kDa
15 kDa
2 kDa
25 kDa
3.5 kDa
50 kDa
8 kDa
Unit Size:
Each
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity MWCO unitSize
11465919 15 mm Each 10 kDa Each
11415879 24 mm Each 1 kDa Each
11425879 45 mm Each 1 kDa Each
11435909 24 mm Each 2 kDa Each
11445909 29 mm Each 2 kDa Each
11425909 11.5 mm Each 2 kDa Each
11455909 11.5 mm Each 3.5 kDa Each
11475909 34.4 mm Each 3.5 kDa Each
11465909 29 mm Each 3.5 kDa Each
11405919 7.5 mm Each 8 kDa Each
11485909 15 mm Each 8 kDa Each
11425919 32 mm Each 8 kDa Each
11415919 20.4 mm Each 8 kDa Each
11495909 25.5 mm Each 8 kDa Each
11455919 7.5 mm Each 10 kDa Each
11475919 20.4 mm Each 10 kDa Each
11485919 29 mm Each 10 kDa Each
11425929 7.5 mm Each 15 kDa Each
11445929 15 mm Each 15 kDa Each
11455949 7.5 mm Each 25 kDa Each
11435949 17.8 mm Each 25 kDa Each
11465929 29 mm Each 15 kDa Each
11445949 21.6 mm Each 25 kDa Each
11455929 20.4 mm Each 15 kDa Each
10749445 7.5 mm Each 50 kDa Each
10071954 17.8 mm Each 50 kDa Each
10031954 21.6 mm Each 50 kDa Each
11435929 3.8 mm 1 Pk. 15 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
This item is not returnable. View return policy
Product Code. 11465919 Supplier Spectrum™ Supplier No. 132118
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 7 Membrane Tubing
MWCO 10 kDa
Material Regenerated Cellulose
Diameter (Metric) 15 mm
Diameter (English) 0.59 in.
Length (English) 16.4 ft.
Length (Metric) 5 m
Color Transparent
Formulation Packaged wet in 0.05% sodium azide
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.