missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits

Product Code. 15340782
Click to view available options
Diameter (Metric):
48 mm
89 mm
Length (Metric):
18 mL/cm vol./length
62 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
This item is not returnable. View return policy

Product Code. 15340782

Brand: Spectrum™ 132 757T1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

5 m of Spectra/Por 5 dialysis tubing and 2 closures

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

  • For reinforced for weight applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry without glycerin (no soaking required)

Specifications

MWCO 12 to 14 kDa
Diameter (Metric) 89 mm
Length (Metric) 62 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.