missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
Brand: Spectrum™ 133264
Code : NEW
Additional Details : Weight : 0.20000kg
Description
- Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
- Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
- Rigidly controlled porosity for better selectivity and separations
- Ultrapure, does not interfere with molecular activity and function
- 3 MWCOs and 2 tubing flat-widths
- Packaged dry with glycerin (soak 15 min. in water)
- Use with pH 2 to 12 and temp 4 to 60°C
Specifications
Spectra/Por Dialysis Tubing | |
Regenerated Cellulose | |
0.25 in. | |
5 m | |
39.2°F to 140°F | |
Each |
8 to 10 kDa | |
6.4 mm | |
16.4 ft. | |
Packaged dry with glycerin (soak 15 min. in water) | |
4°C to 60°C | |
Frequent dialysis applications, desalting, purification, sample preparation |