missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO

Product Code. 11455869
Change view
Click to view available options
Diameter (Metric):
7.5 mm
11.5 mm
15 mm
17.8 mm
20.4 mm
21.6 mm
24 mm
29 mm
31.8 mm
34.4 mm
MWCO:
1 kDa
10 kDa
15 kDa
2 kDa
25 kDa
3.5 kDa
50 kDa
8 kDa
Unit Size:
Each
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) MWCO unitSize
11455869 11.5 mm 2 kDa Each
11425869 11.5 mm 1 kDa Each
11435869 24 mm 1 kDa Each
11445869 29 mm 1 kDa Each
11465869 24 mm 2 kDa Each
11485869 11.5 mm 3.5 kDa Each
11475869 29 mm 2 kDa Each
11405879 34.4 mm 3.5 kDa Each
11445879 15 mm 8 kDa Each
11475879 20.4 mm 8 kDa Each
11495879 31.8 mm 8 kDa Each
11405889 7.5 mm 10 kDa Each
11415889 15 mm 10 kDa Each
11425889 20.4 mm 10 kDa Each
11435889 29 mm 10 kDa Each
11475889 7.5 mm 15 kDa Each
11495889 15 mm 15 kDa Each
11405899 20.4 mm 15 kDa Each
11425899 7.5 mm 25 kDa Each
11435899 17.8 mm 25 kDa Each
11405909 21.6 mm 50 kDa Each
11495899 17.8 mm 50 kDa Each
11485899 7.5 mm 50 kDa Each
11445899 21.6 mm 25 kDa Each
11495869 29 mm 3.5 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
This item is not returnable. View return policy
Product Code. 11455869 Supplier Spectrum™ Supplier No. 132620
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane

This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.

  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 6 Dialysis Tubing
MWCO 2 kDa
Material Regenerated Cellulose
Diameter (Metric) 11.5 mm
Diameter (English) 0.45 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Packaged wet in 0.05% sodium azide
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.