Learn More
Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Description
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.
- Pre-wetted & ready-to-use, no glycerin removal required
- 8 MWCO's ranging from 1 kD - 50 kD
- Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
- Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
- Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
- Economic 10 meters/pkg
Do not tie knots in tubing; use closures
Specifications
Specifications
| Product Type | Spectra/Por 6 Dialysis Tubing |
| MWCO | 50 kDa |
| Material | Regenerated Cellulose |
| Diameter (Metric) | 17.8 mm |
| Diameter (English) | 0.7 in. |
| Length (English) | 33 ft. |
| Length (Metric) | 10 m |
| Formulation | Packaged wet in 0.05% sodium azide |
| Temperature Range (English) | 39.2°F to 249.8°F |
| Temperature Range (Metric) | 4°C to 121°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.