missing translation for 'onlineSavingsMsg'
Learn More

Reagecon™ Spectrophotometric Standards
BIOPHARMA

Product Code. 11754815 Shop All Reagecon Products
Click to view available options
Type:
Optical standard 1.0
Optical standard 2.0
Optical standard 30.0
Optical standard 8.0
Absorbance Range:
0.0058 to 0.094 A
0.012 to 0.02023 A
0.049 to 0.7928 A
0.1929 to 2.6225 A
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 11754815

Brand: Reagecon™ OPS2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Calibrate a spectrophotometer to ensure accurate transmission measurements. Spectrophotometric Standards, Reagecon™ are safe, stable and easy-to-use optical standards with guaranteed accuracy of ±5%. All solutions are NIST traceable and come with a certificate of analysis.

Spectrophotometry is the quantitative measurement of the reflection or transmission properties of a material. Spectrophotometers are used to measure a light beam's intensity as a function of its color (wavelength). These reference standards act as a calibration and control solution, with highly accurate absorbance data at a variety of wavelengths.

  • NIST Traceable—Instruments used to acquire data are NIST calibrated
  • Easy to Use—Pour straight out of the plastic bottle
  • Accurate—Guaranteed accuracy of ±5% inter/intra-instrument variance
  • Stable—Will not settle out of solution, guaranteed one year shelf life
  • Safe—Non carcinogenic materials used

Specifications

Absorbance Range 0.012 to 0.02023 A
Certifications/Compliance NIST traceable
Type Optical standard 2.0
Wavelength Range 450 to 224 nm
For Use With (Application) Calibration of spectrophotometers
Includes Certificate of analysis
Packaging 125 mL Plastic Bottles
Storage Requirements Shelf life: 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.