missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagecon™ Spectrophotometric Standards
Shop All Reagecon Products
Click to view available options
Type:
Optical standard 1.0
Optical standard 2.0
Optical standard 30.0
Optical standard 8.0
Absorbance Range:
0.0058 to 0.094 A
0.012 to 0.02023 A
0.049 to 0.7928 A
0.1929 to 2.6225 A
Unit Size:
125mL
Description
Spectrophotometry is the quantitative measurement of the reflection or transmission properties of a material. Spectrophotometers are used to measure a light beam's intensity as a function of its color (wavelength). These reference standards act as a calibration and control solution, with highly accurate absorbance data at a variety of wavelengths.
- NIST Traceable—Instruments used to acquire data are NIST calibrated
- Easy to Use—Pour straight out of the plastic bottle
- Accurate—Guaranteed accuracy of ±5% inter/intra-instrument variance
- Stable—Will not settle out of solution, guaranteed one year shelf life
- Safe—Non carcinogenic materials used
Specifications
Specifications
| Absorbance Range | 0.0058 to 0.094 A |
| Certifications/Compliance | NIST traceable |
| Type | Optical standard 1.0 |
| Wavelength Range | 450 to 224 nm |
| For Use With (Application) | Calibration of spectrophotometers |
| Includes | Certificate of analysis |
| Packaging | 125 mL Plastic Bottles |
| Storage Requirements | Shelf life: 1 year |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction