Learn More
Spectroflux 105, Lithium tetraborate, Lithium carbonate & Lanthanum oxide, 47:37:16 w/w%
Description
A basic oxidizing flux. Suitable for sulfate, phosphate, and other acidic minerals. Will oxidize traces of reduced species. Used where computer signal processing is limited
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 700°C |
| Physical Form | Solid |
| Quantity | 1 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.