Learn More
Thermo Scientific Alfa Aesar SPECTROFLUX 100B 1KG
Brand: Thermo Scientific Alfa Aesar 012080.A1
SPECTROFLUX 100B 1KG
Description
Safety and Handling
- Spectroflux 100B, Lithium metaborate & Lithium tetraborate, 80:20 w/w%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Lithium metaborate,Lithium tetraborate