missing translation for 'onlineSavingsMsg'
Learn More

Spectroflux 100, low phosphorus, Lithium tetraborate, phosphorus <20ppm

Product Code. 11344937
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344937 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11344937 Supplier Thermo Scientific Alfa Aesar Supplier No. 041951.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A general purpose flux with a low phosphorus content. Dissolves most refractories. Often used in testing of ceramics. Not suitable for highly acidic samples. Samples must be fully oxidized | CAS: 12007-60-2

A general purpose flux with a low phosphorus content. Dissolves most refractories. Often used in testing of ceramics. Not suitable for highly acidic samples. Samples must be fully oxidizedSpectroflux 100, low phosphorus, Lithium tetraborate is used to dissolve refractories. It is a spectroscopic fluxing agent which is utilized for fusing basic oxides, glass, alumino-silicates, ceramics, carbonates, cement, steel, aluminum, soil, concrete mixes, bauxites, magnesites and rare earth oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A general purpose flux with a low phosphorus content. Dissolves most refractories. Often used in testing of ceramics. Not suitable for highly acidic samples. Samples must be fully oxidized
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 920°C
CAS 12007-60-2
Physical Form Solid
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.