Learn More
Specac™ Spectroelectrochemical Transmission Cell
Click to view available options
Type:
Spectroelectrochemical Transmission Cell
Description
This is an FTIR accessory encompassing a think layer optically transparent electrode structure to facilitate electrochemical spectroscopic measurements. The cell comprises two CaF2 windows separated by a 100 or 200 micron sample layer, where rhodium grid working and counter electrodes, and a silver reference electrode, are photolithographically imprinted on the surface of the front window in contact with the sample.
Specifications
Specifications
| Type | Spectroelectrochemical Transmission Cell |
| Includes | Two CaF2 windows separated by a 100 or 200 micron sample layer, where rhodium grid working and counter electrodes, and a silver reference electrode, are photolithographically imprinted on the surface of the front window in contact with the sample |