Learn More
Invitrogen™ Spectrin beta-3 Recombinant Superclonal™ Antibody (14HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710971
Description
This antibody is predicted to react with Monkey, Cat and Rat. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Spectrins are principle components of a cell's membrane-cytoskeleton and are composed of two alpha and two beta spectrin subunits. The protein encoded by this gene (SPTBN2), is called spectrin beta non-erythrocytic 2 or beta-III spectrin. It is related to, but distinct from, the beta-II spectrin gene which is also known as spectrin beta non-erythrocytic 1 (SPTBN1). SPTBN2 regulates the glutamate signaling pathway by stabilizing the glutamate transporter EAAT4 at the surface of the plasma membrane. Mutations in this gene cause a form of spinocerebellar ataxia, SCA5, that is characterized by neurodegeneration, progressive locomotor incoordination, dysarthria, and uncoordinated eye movements.
Specifications
| Spectrin beta-3 | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| O15020 | |
| Sptbn2 | |
| Peptides corresponding to Human SPTBN2 (aa 1334-1351, 1733-1750). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 14HCLC | |
| Unconjugated | |
| Sptbn2 | |
| beta SpIII sigma 1; beta-III spectrin; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; GTRAP41; KIAA0302; mKIAA0302; SCA5; SCAR14; Spectrin Beta; spectrin beta 3; spectrin beta chain, brain 2; spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin beta, non-erythrocytic 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; spectrin-like protein GTRAP41; spinocerebellar ataxia 5 protein; Spnb3; SPNB-3; SPTBN2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.