missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 3 Rabbit anti-Human, Clone: SPTBN2/2887R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07293-100ug
This item is not returnable.
View return policy
Description
Spectrin beta 3 Monoclonal specifically detects Spectrin beta 3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Spectrin beta 3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5 | |
| Recombinant human Spectrin beta 3 fragment (aa356-475) (exact sequence is proprietary) | |
| 100 μg | |
| Neuroscience | |
| 6712 | |
| Store at 4°C. | |
| IgG |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPTBN2/2887R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction