missing translation for 'onlineSavingsMsg'
Learn More

Spectrin beta 3 Antibody (SPTBN2/1583), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18408593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408593 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408593 Supplier Novus Biologicals Supplier No. NBP254440AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Spectrin beta 3 Monoclonal antibody specifically detects Spectrin beta 3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spectrin beta 3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPTBN2/1583
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Beta-III spectrin, glutamate transporter EAAT4-associated protein 41, GTRAP41, KIAA0302, SCA5, spectrin beta chain, brain 2, spectrin, beta, non-erythrocytic 2, Spectrin, non-erythroid beta chain 2, spinocerebellar ataxia 5
Host Species Mouse
Immunogen Recombinant fragment (around aa356-475) of human Spectrin beta 3 protein (exact sequence is proprietary) (Uniprot: O15020)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6712
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.