missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93443-0.02ml
This item is not returnable.
View return policy
Description
Spectrin beta 2 Polyclonal antibody specifically detects Spectrin beta 2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Spectrin beta 2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:3000 | |
| beta-fodrin, beta-G spectrin, Beta-II spectrin, beta-spectrin 2, beta-spectrin II, betaSpII, ELF, embryonic liver beta-fodrin, Fodrin beta chain, spectrin beta chain, brain 1, spectrin, beta, non-erythrocytic 1, Spectrin, non-erythroid beta chain 1, SPTB2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2100-2200 of human SPTBN1 (NP_003119.2). PPSPEPSTKVSEEAESQQQWDTSKGEQVSQNGLPAEQGSPRMAETVDTSEMVNGATEQRTSSKESSPIPSPTSDRKAKTALPAQSAATLPARTQETPSAQM | |
| 0.02 mL | |
| Neuroscience | |
| 6711 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction