missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 1 Mouse anti-Human, Mouse, Rat, Clone: RG/26, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Spectrin beta 1 Monoclonal specifically detects Spectrin beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Spectrin beta 1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | RG/26 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | beta-I spectrin, EL3, erythrocyte, HS2, HSPTB1, membrane cytoskeletal protein, spectrin, beta, erythrocytic, SPTB1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?