missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin alpha 1 Rabbit anti-Human, Clone: SPTA1/2939R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07265-100ug
This item is not returnable.
View return policy
Description
Spectrin alpha 1 Monoclonal specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications
| Spectrin alpha 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, ELISA 2 to 4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
| Recombinant fragment of human Spectrin alpha 1 (around aa356-475) (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 6708 | |
| Store at 4°C. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| SPTA1/2939R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction