missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin alpha 1 Mouse anti-Human, Clone: rSPTA1/1832, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00
Specifications
| Antigen | Spectrin alpha 1 |
|---|---|
| Clone | rSPTA1/1832 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Spectrin alpha 1 Monoclonal specifically detects Spectrin alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Spectrin alpha 1 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 6708 | |
| IgG1 κ | |
| Protein A or G purified |
| rSPTA1/1832 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha-I spectrin, EL2, erythrocyte, HS3, spectrin, alpha, erythrocytic 1 (elliptocytosis 2) | |
| Recombinant fragment of human Spectrin alpha 1 (around aa356-475) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title