missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 7 Pre-treated Dialysis Tubing

Product Code. 11485949
Click to view available options
MWCO:
10 kDa
25 kDa
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11485949

Brand: Spectrum™ Labs 128518

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Convenient dispenser boxes protect unused membranes

Pure regenerated natural cellulose membranes with 1kDa to 50kDa MWCO. Transparent and flexible except Spectra/Por™ 5.Low molecular weight compounds in aqueous solutions diffuse through the membrane, higher molecular weights, such as proteins and bacteria, are retained by the membrane. Convenient dispenser boxes protect unused membranes. Glycerol (where used) preserves membrane and must be removed before use. Soak membrane for three hours in deionised water, then rinse well with running water. Spectra/Por™ 5 is reinforced with a layer of porous paper, to give a thick, rigid membrane for use in high shear environments.
  • Spectra/Por™ 1, 2, 3, 4, 5, supplied dry with glycerol
  • Spectra/Por™ 6, pre-wetted supplied in 0.1% sodium azide
  • Spectra/Por™ 7, pre-treated to reduce trace contaminants supplied in 0.1% sodium azide For molecular separation, protein binding studies, electrolyte removal and desalting.

Specifications

Product Type Dialysis Tubing
MWCO 25 kDa
Diameter (Metric) 11.5 mm
Length (Metric) 5 m
Capacity (Metric) 1.1 mL/cm
Quantity 1 Each
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.