Learn More
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs, 12-14kD MWCO
Description
- Dry Standard RC dialysis membrane in single-layer discs.
- Used in various commercially available dialysis equipment.
- Discs are separated by gylcine paper
- Packaged dry with glycerin (rinse out with water)
Specifications
Specifications
| Product Type | Dialysis Membrane Discs |
| MWCO | 12 to 14 kDa |
| Diameter (Metric) | 33 mm |
| Quantity | 50/Pk. |
| For Use With (Application) | Basic dialysis |