Learn More
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs, 12-14kD MWCO
Product Code. 11561010
Click to view available options
Diameter (Metric):
33 mm
47 mm
Unit Size:
Each
Pack of 50
Product Code. 11561010
Brand: Spectrum Labs™ 132 496
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spectrum Labs™ Spectra/Por 4 Dialysis Membrane Discs are for basic dialysis and are hydrophilic with symmetric porosity.
  • Dry Standard RC dialysis membrane in single-layer discs.
  • Used in various commercially available dialysis equipment.
  • Discs are separated by gylcine paper
  • Packaged dry with glycerin (rinse out with water)
Specifications
Product Type Dialysis Membrane Discs
MWCO 12 to 14 kDa
Diameter (Metric) 47 mm
Quantity 50/Pk.
For Use With (Application) Basic dialysis