Learn More
Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder
Shop All Thermo Scientific ProductsDescription
Thermo Scientific Spectra Multicolor Broad Range Protein Ladder is a 4-color (blue, orange, green, pink) protein standard containing 10 prestained proteins (10 to 260 kDa) for use in gel electrophoresis and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications
Specifications
| Content And Storage | Contents: eight vials of 250 μL each |
| Detection Method | Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm) |
| Number of Markers | 10 |
| Ready to Load | Yes |
| Size Range | 10 to 260 kDa |
| Gel Compatibility | Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels |
| Molecular Weight (g/mol) | 260, 140, 100, 70, 50, 40, 35, 25, 15, 10 kDa |
| Quantity | 8 x 250 μL |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Product Line | Spectra |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.