missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder

Product Code. 11862124 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
10 x 250 μL
Unit Size:
Pack of 10
Pack of 2
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11862124 10 x 250 μL Pack of 10
11842124 2 x 250 μL Pack of 2
17136108 8 x 250 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11862124 Supplier Thermo Scientific™ Supplier No. 26623

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Thermo Scientific™ Spectra Multicolor Broad Range Protein Ladder is a 4-color protein standard containing 10 prestained proteins (10 to 260 kDa) for use as size standards in gel electrophoresis and Western blotting.

Thermo Scientific Spectra Multicolor Broad Range Protein Ladder is a 4-color (blue, orange, green, pink) protein standard containing 10 prestained proteins (10 to 260 kDa) for use in gel electrophoresis and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Contents: 10 vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

Detection Method Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Number of Markers 10
Ready to Load Yes
Size Range 10 to 260 kDa
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 260, 140, 100, 70, 50, 40, 35, 25, 15, 10 kDa
Quantity 10 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Line Spectra
Product Type Protein Ladder
Stain Type Blue, Orange, Green, Pink
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.