missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Sulfate, ≥98%, MP Biomedicals™

Product Code. 15563457 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15563457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15563457 Supplier MP Biomedicals Supplier No. 0215899305
  • ONLINE EXCLUSIVE PRICE
    £145.00
    £135.45 / 5g
    Save £9.55
    6% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23312-56-3
Molecular Formula C14H26N2O11S
Molecular Weight (g/mol) 430.43
MDL Number MFCD00270185
InChI Key XGBFWQUQYQIFLB-MTTMTQIXSA-N
Synonym U 18409E, antibiotic
ChEBI CHEBI:9216
IUPAC Name (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
SMILES OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Type Spectinomycin Sulfate
Physical Form Powder
Color White
Quantity 5 g
Assay Percent Range ≥98%
PubChem CID 64771
Formula Weight 430.4
Percent Purity ≥98%
Grade Not Available
Chemical Name or Material Spectinomycin Sulfate
compliance-icons compliance-icons
Product Identifier
  • Spectinomycin Sulfate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local, regional, national and international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.