missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin sulfate, 93%

Product Code. 15467229
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15467229 1 g Glass bottle 1g
15477229 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467229 Supplier Thermo Scientific Alfa Aesar Supplier No. J66034.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23312-56-3 | C14H26N2O11S | 430.43 g/mol

Targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23312-56-3
Molecular Formula C14H26N2O11S
Molecular Weight (g/mol) 430.43
MDL Number MFCD00270185
InChI Key XGBFWQUQYQIFLB-MTTMTQIXSA-N
Synonym spectinomycin sulfate, spectinomycin sulphate, unii-bz0h4tlf9x, actinospectacin, trobicin, bz0h4tlf9x, togamycin sulfate 1:1, spectinomycin sulfate hydrate, spectinomicina, spectogard
PubChem CID 64771
IUPAC Name (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
SMILES OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Boiling Point 583°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 1 g
Solubility Information Soluble in DMSO or water
Formula Weight 430.43
Percent Purity 93%
Physical Form Powder
Chemical Name or Material Spectinomycin sulfate
compliance-icons
Product Identifier
  • Spectinomycin sulfate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Spectinomycin sulphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.