missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Dihydrochloride Pentahydrate, TRC

Product Code. 30424717
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424717 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424717 Supplier Toronto Research Chemicals Supplier No. TRCS6802585G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spectinomycin dihydrochloride pentahydrate
CAS 22189-32-8
Molecular Formula C14 H24 N2 O7 . 2 Cl H . 5 H2 O
Quantity 5 g
Synonym 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one','decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-','dihydrochloride','pentahydrate (8CI), 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one','decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-','dihydrochloride','pentahydrate','(2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)- (9CI), 4H-Pyrano[2,3-b][1,4]benzodioxin-4-one','decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-','dihydrochloride','pentahydrate','[2R-(2α,4aβ,5aβ,6β,7β,8β,9α,9aα,10aβ)]-, Spectinomycin dihydrochloride pentahydrate, Spectinomycin hydrochloride hydrate
InChI Formula InChI=1S/C14H24N2O7.2ClH.5H2O/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;;;;;;/h5,7-13,15-16,18-20H,4H2,1-3H3;2*1H;5*1H2/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;;;;;;/m1......./s1
SMILES O.O.O.O.O.Cl.Cl.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
IUPAC Name (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-Decahydro-4a,7,9-trihydroxy-2-methyl-6,8-bis(methylamino)-4H-Pyrano[2,3-b][1,4]benzodioxin-4-one dihydrochloride pentahydrate
Molecular Weight (g/mol) 495.3478
Formula Weight 494.1645
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.