missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specpure™ Acetate, Ion Chromatography Standard Solution, Alfa Aesar™
Guaranteed stable for six months from the ship date; keep refrigerated
Brand: Alfa Aesar 042563.AP
Code : NEW
Additional Details : Weight : 0.01000kg
Quantity | 500mL |
---|
Chemical Identifiers
Acetate | |
Liquid | |
MFCD00282007 |
CH3CO2? 1000μg/mL | |
CH3CO2Na in H2O |
Specifications
Acetate | |
Clear liquid | |
Liquid | |
MFCD00282007 | |
500mL |
CH3CO2? 1000μg/mL | |
Odorless | |
CH3CO2Na in H2O | |
Clear colorless |