Learn More
BINDER™ Specimen Temperature Display
Product Code. p-200104340 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO Series
KB Series
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238262
Brand: BINDER™ 80122549
Only null left
Add to basket
Add to basket
Flexible Pt100 indoor temperature sensor
Platinum 100 ohm temperature sensor for sample temperature display or control.
Specifications
For Use With (Equipment) KB Series
Material Steel
Type Temperature Sensor