missing translation for 'onlineSavingsMsg'
Learn More

Specac™ Specamill™ Grinding Accessory 220V UK

Product Code. 15220048
Encompass_Preferred
Click to view available options
Volume (Metric):
3 mL
This item is not returnable. View return policy

Product Code. 15220048

Brand: Specac™ GS06000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
The Specamill with UK plug is a grinding device designed for the preparation of samples prior to analysis. Can be supplied with 2 grinding sets, stainless steel capsule sets or agate capsule sets. Stainless Steel is a better choice for harder materials. The capacity of each capsule is 3ml and the Mill features a timer with adjustable vibration.

Specifications

Includes 2 grinding sets, stainless steel capsule sets or agate capsule sets
Volume (Metric) 3 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.