Learn More
Specac™ Specadie™ Kit Complete
Description
This kit is made up of a Specadie, Holder, 2 x Wrenches, Spare set of bolts and seals and pot of KBr Powder. Designed for pellet production without the need for a press and die.
Specifications
Specifications
| Type | Specadie™ Kit Complete |
| Includes | Specadie, Holder, 2 x Wrenches, Spare set of bolts and seals and pot of KBr Powder |