missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Specacards for FTIR 10mm x 25mm Aperture
Click to view available options
Diameter (Metric):
10 mm
Description
These Specacards are designed to hold a sample post production to be directly mounted into an FTIR instrument for analysis. This product has a rectangular aperture of 10mm diameter.
Specifications
Specifications
| Diameter (Metric) | 10 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction