missing translation for 'onlineSavingsMsg'
Learn More

SPE tube ODS polar plus reversed phase polypropylene 3g, 6mL Bakerbond

Brand:  JT Baker 7466-10

Code : Z3

Additional Details : CAS Number : 71889-02-6 Weight : 0.10000kg

Product Code. 13111634

  • POA / Pack of 30
Estimated Shipment: 12-06-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.