Learn More
Macherey-Nagel™ SPE Column Kit
Product Code. 10117361 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Application):
SPE Analysis of PCBs
Unit Size:
Each
Product Code. For Use With (Application) unitSize
10117361 SPE Analysis of PCBs Each
1 options
Product Code. 10117361 Supplier Macherey-Nagel™ Supplier No. 730125
Specifications
Column Type SPE
For Use With (Application) SPE Analysis of PCBs
Type Solid Phase Extration Column Kit
Includes 25 columns each of Chromabond SiOH-H2 SO4/SA and Chromabond SiOH