Learn More
Gerstel µSPE Cartridge LC QuEChERS Sorbent Mix 30mg C18EC(21mg ),Z-Sep(8mg),GCB(1mg ) 96 cartridges
Brand:  Gerstel 100100-602-00
Encompass_Preferred
µSPE Cartridge LC QuEChERS Sorbent Mix 30mg C18EC(21mg ),Z-Sep(8mg),GCB(1mg ) 96 cartridges
Product Code. 30216911
  • £735.62 / Each
Only null left
Estimated Shipment: 07-01-2026