Learn More
Invitrogen™ SPDYA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596075
Description
Immunogen sequence: MRHNQMCCET PPTVTVYVKS GSNRSHQPKK PITLKRPICK DNWQAFEKNT HNNNKSKRPK GPCLVIQRQD MTAFFKLFDD DLIQDFLWMD CCCKIADKYL LAMTFVYFKR AKFTISEHTR INFFIALYLA NTVEEDEEET KYEIFPWALG KNWRKLFPNF LKLRDQLWDR IDYRAIVSRR CCEEVMAIAP THYIWQRERS VHHSGAVRNY NRDEVQLPRG PSATPVDCSL CGKKRRYVRL GLSSSSSLSS HTAGVTEKHS QDSYNSLSMD IIGDPSQAYT GSEVVNDHQS NKGKKTNFLK KDKSMEWFTG SEE; Positive Samples: A-549, Mouse ovary, Rat testis; Cellular Location: Nucleus.
Speedy A, also known as SPDYA, SPDY1, Ringo3 or SPY1, is a 313 amino acid protein that localizes to the nucleus and belongs to the speedy/ringo family. Expressed at high levels in testis and at lower levels in brain, kidney, heart, bone marrow, colon, lung, liver spleen and placenta, Speedy A functions to regulate the G1/S phase transition of the cell cycle, specifically by binding to and activating Cdc2 p34, Cdk2 and p27. Additionally, Speedy A mediates cell survival during DNA damage, suggesting that Speedy A plays a role in proper cell cycle progression throughout the life of the cell. Multiple isoforms of Speedy A exist due to alternative splicing events. The gene encoding Speedy A maps to human chromosome 2, which encodes over 1,400 genes and comprises nearly 8% of the human genome.
Specifications
| SPDYA | |
| Polyclonal | |
| Unconjugated | |
| SPDYA | |
| 4921517J08Rik; 4930548B21Rik; Gs4; hSpy/Ringo A; Lm23; MLZ-465; mSpy/Ringo A; protein expressed in male leptotene and zygotene spermatocytes 465; Rapid inducer of G2/M progression in oocytes A; RINGO A; RINGO3; RINGOA; SPDY1; SPDYA; spdya {ECO:0000250; speedy A; speedy A1; speedy A2; speedy homolog 1; speedy homolog A; speedy homolog A (Xenopus laevis); speedy protein A; speedy/ringo; speedy/RINGO cell cycle regulator family member A; Speedy-1; spermatogenesis related protein Gs4; spy1; UniProtKB:Q5MJ70} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192209, 245711, 70891 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5IBH7, Q5MJ70, Q8R496 | |
| SPDYA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-313 of human SPDYA (NP_8774332). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.