missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPDP, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Description
SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).
Specifications
Specifications
| Chemical Name or Material | SPDP |
| CAS | 68181-17-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H12N2O4S2 |
| Quantity | 50 mg |
| Synonym | SPDP Crosslinker |
| Solubility Information | DMSO : 155 mg/mL (496.22 mM; Need ultrasonic) |
| SMILES | O=C(ON1C(CCC1=O)=O)CCSSC2=NC=CC=C2 |
| Molecular Weight (g/mol) | 312.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction