missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPDEF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94491-0.02ml
This item is not returnable.
View return policy
Description
SPDEF Polyclonal antibody specifically detects SPDEF in Human samples. It is validated for Western Blot
Specifications
| SPDEF | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bA375E1.3, PDEFRP11-375E1__A.3, Prostate epithelium-specific Ets transcription factor, Prostate-derived Ets factor, Prostate-specific Ets, PSE, SAM pointed domain containing ets transcription factor, SAM pointed domain-containing Ets transcription factor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-335 of human SPDEF (NP_036523.1). MGSASPGLSSVSPSHLLLPPDTVSRTGLEKAAAGAVGLERRDWSPSPPATPEQGLSAFYLSYFDMLYPEDSSWAAKAPGASSREEPPEEPEQCPVIDSQAPAGSLDLVPGGLTLEEHSLEQVQSMVVGEVLKDIETACKLLNITADPMDWSPSNVQKWLLWTEHQYRLPPMGKAFQELAGKELCAMSEEQFRQRSPLGGDVLHAHLDIWKSAAWMKERTSPGAIHYCASTSEESWTDSEVDSSCSGQPIHLWQFLKELLLKPHSYGRFIRWLNKEKGIFKIEDSAQVARLWGIRKNRPAMNYDKLSRSIRQYYKKGIIRKPDISQRLVYQFVHPI | |
| 0.02 mL | |
| Cancer | |
| 25803 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction